Skip to Content
Merck
All Photos(1)

Key Documents

374482

Sigma-Aldrich

N-Benzylquininium chloride

95%

Synonym(s):

QUIBEC

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31ClN2O2
CAS Number:
Molecular Weight:
451.00
Beilstein:
5702637
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

optical activity

[α]20/D −235°, c = 1.5 in H2O

mp

200-205 °C (dec.) (lit.)

functional group

hydroxyl
phenyl

SMILES string

[Cl-].COc1ccc2nccc([C@@H](O)[C@@H]3C[C@@H]4CC[N@+]3(C[C@@H]4C=C)Cc5ccccc5)c2c1

InChI

1S/C27H31N2O2.ClH/c1-3-20-18-29(17-19-7-5-4-6-8-19)14-12-21(20)15-26(29)27(30)23-11-13-28-25-10-9-22(31-2)16-24(23)25;/h3-11,13,16,20-21,26-27,30H,1,12,14-15,17-18H2,2H3;1H/q+1;/p-1/t20-,21-,26-,27+,29+;/m0./s1

InChI key

JYDIJFKNXHPWBJ-GOGFHWEMSA-M

Looking for similar products? Visit Product Comparison Guide

General description

N-Benzylquininium chloride (or Quibec) belongs to the class of cinchona family of alkaloids. It is used as a catalyst in the presence of hydroxide bases in various phase transfer reactions, epoxidations, alkylations, and Michael reactions.

Application

N-Benzylquininium chloride can be used as a phase transfer catalyst:
  • In the sulfenylation of various β-keto sulfoxides.
  • To synthesize N-carbamoyl-protected β-nitroamines from α-amido sulfones by asymmetric aza-Henry reaction.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sulfenylation of β-keto sulfoxides. III. Diastereoselectivity induced by a chiral phase transfer catalyst
Wladislaw B, et al.
Tetrahedron, 55(41), 12023-12030 (1999)
N-Benzylquininium Chloride
Bos Mary E, et al.
Encyclopedia of Reagents for Organic Synthesis, Second Edition, 1-7 (2001)
Phase-Transfer-Catalyzed Asymmetric Aza-Henry Reaction Using N-Carbamoyl Imines Generated In Situ from ?-Amido Sulfones
Fini F, et al.
Angewandte Chemie (International ed. in English), 44(48), 7975-7978 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service