Skip to Content
Merck
All Photos(1)

Key Documents

374431

Sigma-Aldrich

Poly(perfluoropropylene oxide-co-perfluoroformaldehyde)

average Mn ~3,200

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CF(CF3)CF2O]x(CF2O)y
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

feed ratio

30:1 (x:y)

mol wt

average Mn ~3,200

refractive index

n20/D 1.3

density

1.888 g/mL at 25 °C

SMILES string

OC(O)(F)F.OC(F)(F)C(O)(F)C(F)(F)F

Looking for similar products? Visit Product Comparison Guide

Application

High performance lubricant for textile, bearing, and corrosive environment applications.

Features and Benefits

Polymer is made by an oxidative polymerization of hexafluoropropylene and tetrafluoroethylene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

>235.4 °F - closed cup

Flash Point(C)

> 113.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service