Skip to Content
Merck
All Photos(1)

Key Documents

374377

Sigma-Aldrich

2-(Triphenylphosphoranylidene)propionaldehyde

98%

Synonym(s):

1-Formylethylidenetriphenylphosphorane

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$329.00

$329.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$329.00

About This Item

Linear Formula:
(C6H5)3P=C(CH3)CHO
CAS Number:
Molecular Weight:
318.35
MDL number:
UNSPSC Code:
12352114
PubChem Substance ID:
NACRES:
NA.22

$329.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

reaction suitability

reaction type: C-C Bond Formation

mp

219-221 °C (lit.)

functional group

aldehyde
phosphine

SMILES string

CC(C=O)=P(c1ccccc1)(c2ccccc2)c3ccccc3

InChI

1S/C21H19OP/c1-18(17-22)23(19-11-5-2-6-12-19,20-13-7-3-8-14-20)21-15-9-4-10-16-21/h2-17H,1H3

InChI key

VHUQEFAWBCDBSC-UHFFFAOYSA-N

Application

Reactant for:
  • Preparation of acitretin analogs with antitumor activity
  • Stereoselective synthesis of essential building blocks of phorboxazole A via hetero Diels-Alder and stereoselective hetero-Michael addition/equilibration
  • Preparation of C1-C14 fragment of sarcoglaucol-16-one via Z-selective ando-type Horner-Wadsworth-Emmons olefination
  • Wittig reactions
  • Synthesis of ring-expanded and epothilone-hybridized macrosphelide analogs as apoptosis-inducing agents
  • Preparation of phosphonic acid-containing analogs of mycophenolic acid as inhibitors of IMPDH

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service