Skip to Content
Merck
All Photos(1)

Key Documents

374024

Sigma-Aldrich

3-Methoxythiophene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$166.00

$166.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$166.00

About This Item

Empirical Formula (Hill Notation):
C5H6OS
CAS Number:
Molecular Weight:
114.17
Beilstein:
106404
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$166.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.532 (lit.)

bp

80-82 °C/65 mmHg (lit.)

density

1.143 g/mL at 25 °C (lit.)

SMILES string

COc1ccsc1

InChI

1S/C5H6OS/c1-6-5-2-3-7-4-5/h2-4H,1H3

InChI key

RFSKGCVUDQRZSD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methoxythiophene is a thiophene. The intramolecular and intermolecular geometries of crystals of 3-methoxythiophene were studied.[1] Spectroscopic studies of bipolarons derived from oligomerized 3-methoxythiophene in solution has been reported.[2] The electropolymerization of 3-methoxythiophene on Pt and Fe electrodes in an aqueous micellar medium containing sodium dodecyl sulfate and 10-3M bithiophene has been reported.[3] Thin polymer films of 3-methoxythiophene at the cathode in a direct current discharge have been prepared.[4]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

120.2 °F - closed cup

Flash Point(C)

49 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paolo Bollella et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 21(1), 120-128 (2019-08-14)
Biocatalytic buckypaper electrodes modified with pyrroloquinoline quinone (PQQ)-dependent glucose dehydrogenase and bilirubin oxidase for glucose oxidation and oxygen reduction, respectively, were prepared for their use in a biofuel cell. A small (millimeter-scale; 2×3×2 mm3 ) enzyme-based biofuel cell was tested in
Martina Marinelli et al.
Chirality, 32(12), 1361-1376 (2020-11-17)
Novel optically active oligothiophenes bearing electron-donating chiral side chains have been prepared by synthetic methods suitable to achieve regioregular head-to-tail and head-to-head/tail-to-tail derivatives. In particular, the chiral (S)-(2-methyl)butyl moiety was linked at position 3 of the thiophene ring through heteroatoms
Electrosynthesis and characterization of poly (3-methoxythiophene)-polybithiophene composite films prepared in micellar media on Pt and Fe substrates.
Dieng MaM, et al.
Physical Chemistry Chemical Physics, 1(8), 1731-1734 (1999)
Spectroscopic studies of bipolarons from oligomerized 3-methoxythiophene in solution.
Chang A-C and Miller LL.
Synthetic Metals, 22(1), 71-78 (1987)
Blake et al.
Acta crystallographica. Section B, Structural science, 55(Pt 6), 963-974 (2000-08-06)
The intramolecular and intermolecular geometries of six thiophenes carrying oxygen-containing substituents have been determined. Crystals of 2-methoxythiophene and 3-methoxythiophene were grown in situ on a diffractometer from liquid samples. The 2-methoxy group introduces significant distortions to the thiophene nucleus and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service