Skip to Content
Merck
All Photos(1)

Key Documents

371998

Sigma-Aldrich

2,2,3,3-Tetrafluoropropyl methacrylate

99%

Synonym(s):

1H ,1H ,3H -Tetrafluoropropyl methacrylate, 2,2,3,3-Tetrafluoropropyl 2-methylprop-2-enoate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$287.00

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$287.00

About This Item

Linear Formula:
CH2=C(CH3)COOCH2CF2CF2H
CAS Number:
Molecular Weight:
200.13
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$287.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

form

liquid

contains

BHT as stabilizer

refractive index

n20/D 1.373 (lit.)

bp

124 °C (lit.)

density

1.25 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCC(F)(F)C(F)F

InChI

1S/C7H8F4O2/c1-4(2)5(12)13-3-7(10,11)6(8)9/h6H,1,3H2,2H3

InChI key

RSVZYSKAPMBSMY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

123.8 °F - closed cup

Flash Point(C)

51 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service