Skip to Content
Merck
All Photos(1)

Key Documents

370215

Sigma-Aldrich

Methyl 3,5-dimethylbenzoate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$35.30

$35.30


Estimated to ship onApril 11, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$35.30

About This Item

Linear Formula:
(CH3)2C6H3CO2CH3
CAS Number:
Molecular Weight:
164.20
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$35.30


Estimated to ship onApril 11, 2025


Request a Bulk Order

Quality Level

Assay

98%

form

solid

bp

239-240 °C (lit.)

mp

31-33 °C (lit.)

density

1.027 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

COC(=O)c1cc(C)cc(C)c1

InChI

1S/C10H12O2/c1-7-4-8(2)6-9(5-7)10(11)12-3/h4-6H,1-3H3

InChI key

PEVXENGLERTHJE-UHFFFAOYSA-N

General description

Methyl 3,5-dimethylbenzoate is an aromatic carboxylic acid ester. It was selected as ligand during monomer screening for the synthesis and investigation of various europium compounds containing pinacolyl methylphosphonate with different ligands.[1] Methyl 3,5-dimethylbenzoate is reported as precursor of methyl-3,5-divinylbenzoate.[2]

Application

Methyl 3,5-dimethylbenzoate may be used in the preparation of four carbon isostere related to highly active 4-pyridinemethanols, which were subsequently evaluated for their antimalarial activity.[3] It may be used in the total synthesis of (±)-indoxamycin B.[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107.00 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecularly imprinted polymers for the selective sequestering and sensing of ions.
The Johns Hopkins Medical Letter Health After 50, 18(4), 465-465 (1997)
Oliver F Jeker et al.
Angewandte Chemie (International ed. in English), 51(14), 3474-3477 (2012-02-22)
Revised version: the first total synthesis of indoxamycin B leads to a stereochemical reassignment of the natural product. The synthetic route features an efficient carboannulation sequence to rapidly access the dihydroindenone system. Moreover, a series of Au(I)-catalyzed transformations served in
A Markovac et al.
Journal of medicinal chemistry, 23(11), 1198-1201 (1980-11-01)
Four carbon isosteres related to the highly active 4-pyridylcarbinolamines were prepared and evaluated for suppressive antimalarial activity against Plasmodium berghei in mice. Three of the four examples possessed significant activity but were approximately one dose level less active than the
A L Jenkins et al.
Analytical chemistry, 71(2), 373-378 (1999-02-09)
The techniques of molecular imprinting and sensitized lanthanide luminescence have been combined to create the basis for a sensor that can selectively measure the hydrolysis product of the nerve agent Soman in water. The sensor functions by selectively and reversibly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service