Skip to Content
Merck
All Photos(1)

Key Documents

369098

Sigma-Aldrich

3-Chloro-4-fluorobenzoic acid

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)CO2H
CAS Number:
Molecular Weight:
174.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

95%

mp

133-135 °C (lit.)

SMILES string

OC(=O)c1ccc(F)c(Cl)c1

InChI

1S/C7H4ClFO2/c8-5-3-4(7(10)11)1-2-6(5)9/h1-3H,(H,10,11)

InChI key

PKTSBFXIHLYGEY-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Herbicidal Activity of 3-Chloro-4-fluorobenzoylthiourea [J].
Chang-chun LIU.
Chemical World, 3, 010-010 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service