Skip to Content
Merck
All Photos(1)

Key Documents

367508

Sigma-Aldrich

7-Methoxy-2-naphthol

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$86.10

$86.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$86.10

About This Item

Linear Formula:
CH3OC10H6OH
CAS Number:
Molecular Weight:
174.20
Beilstein:
2207279
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$86.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

mp

116-119 °C (lit.)

SMILES string

COc1ccc2ccc(O)cc2c1

InChI

1S/C11H10O2/c1-13-11-5-3-8-2-4-10(12)6-9(8)7-11/h2-7,12H,1H3

InChI key

UNFNRIIETORURP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

7-Methoxy-2-naphthol is a 7-substituted-2-naphthol.[1] Crystal structure of 7-methoxy-2-naphthol has been reported.[2] Reaction of 7-methoxy-2-naphthol with N-methyl-N-phenylhydrazine under thermal conditions has been reported.[3]

Application

7-Methoxy-2-naphthol may be used in the synthesis of new 1H-benzo[f]chromene derivatives.[1] It may be used in the synthesis of 7-methoxy-2-naphthyl (MN)-derived xylosides in which the methoxy group served as a marker for NMR characterization and UV detection.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of 9-methoxy and 9-acetoxy-3-amino-1-(4-methoxyphenyl)-1H-benzo [f] chromene-2-carbonitriles via 2-(imino-piperidin-1-yl-methyl)-3-(4-methoxyphenyl) acrylonitrile as intermediate.
El-Agrody AM, et al.
Zeitschrift fur Naturforschung, B: Chemical Sciences, 57(5), 579-585 (2002)
Sandrine Gulberti et al.
The Journal of biological chemistry, 280(2), 1417-1425 (2004-11-04)
We determined whether the two major structural modifications, i.e. phosphorylation and sulfation of the glycosaminoglycan-protein linkage region (GlcAbeta1-3Galbeta1-3Galbeta1-4Xylbeta1), govern the specificity of the glycosyltransferases responsible for the biosynthesis of the tetrasaccharide primer. We analyzed the influence of C-2 phosphorylation of
7-Methoxy-2-naphthol.
Prince P, et al.
Acta Crystallographica Section C, Crystal Structure Communications, 47(10), 2218-2220 (1991)
Qiang Tang et al.
Journal of the American Chemical Society, 130(18), 5840-5841 (2008-04-15)
An unexpected reaction of N,N-disubstituted hydrazine with naphthol and its analogues under simply thermal conditions has been disclosed. 2-Naphthol reacted with various N,N-disubstituted hydrazines under argon to afford 1-amino-2-naphthol and the corresponding secondary amines in excellent yields. Ortho amination of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service