Skip to Content
Merck
All Photos(1)

Key Documents

366617

Sigma-Aldrich

Vanadium(IV) chloride

Synonym(s):

Vanadium tetrachloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
VCl4
CAS Number:
Molecular Weight:
192.75
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:

form

liquid

reaction suitability

reagent type: catalyst
core: vanadium

bp

154 °C (lit.)

mp

−28 °C (lit.)

solubility

diethyl ether: slightly soluble(lit.)
ethanol: slightly soluble(lit.)

density

1.816 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

Cl[V](Cl)(Cl)Cl

InChI

1S/4ClH.V/h4*1H;/q;;;;+4/p-4

InChI key

JTJFQBNJBPPZRI-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuhan Wang et al.
Journal of materials chemistry. B, 8(27), 5883-5891 (2020-06-17)
It remains a great challenge to integrate effective photothermal therapeutic materials with upconversion nanoparticles (UCNPs) into one structure with small size. Herein, a new and simple method was developed to combine the luminescent UCNPs with vanadium disulfide (VS2) heterogeneously growing

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service