Skip to Content
Merck
All Photos(2)

Documents

365440

Sigma-Aldrich

2-Hydroxy-4-methylpyridine

99%

Synonym(s):

4-Methyl-2(1H)-pyridinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7NO
CAS Number:
Molecular Weight:
109.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

bp

186-187 °C/12 mmHg (lit.)

mp

131-134 °C (lit.)

SMILES string

Cc1ccnc(O)c1

InChI

1S/C6H7NO/c1-5-2-3-7-6(8)4-5/h2-4H,1H3,(H,7,8)

InChI key

YBDRFJXGJQULGH-UHFFFAOYSA-N

General description

2-Hydroxy-4-methylpyridine is formed by the tautomerization of 4-methyl pyridine during the synthesis of bis aldehydes.

Application

2-Hydroxy-4-methylpyridine is suitable for use in a study to investigate the equilibrium of self-association of 2-pyrrolidone (A) and 2-hydroxyl-4-methyl-pyridine (B), also the hetero-association between A and B in [2H3] acetonitrile by NMR.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NMR Study of Hetero-Association Accompanying Self-Association of 2-Hydroxyl-4-methyl-pyridine and 2-Pyrrolidone in Acetonitrile.
Luo W-C, et al.
J. Chin. Chem. Soc., 47(6), 1177-1183 (2000)
Total synthesis of the ocular age pigment A2-E: a convergent pathway.
Ren R X-F, et al.
Journal of the American Chemical Society, 119(15), 3619-3620 (1997)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service