Skip to Content
Merck
All Photos(1)

Key Documents

362719

Sigma-Aldrich

Di-tert-butylphosphine

98%

Synonym(s):

Bis(tert-butyl)phosphine, HP(t-Bu)2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CPHC(CH3)3
CAS Number:
Molecular Weight:
146.21
Beilstein:
1918492
MDL number:
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

reaction suitability

reagent type: ligand
reaction type: Buchwald-Hartwig Cross Coupling Reaction

reagent type: ligand
reaction type: Heck Reaction

reagent type: ligand
reaction type: Suzuki-Miyaura Coupling

density

0.79 g/mL at 25 °C (lit.)

functional group

phosphine

SMILES string

CC(C)(C)PC(C)(C)C

InChI

1S/C8H19P/c1-7(2,3)9-8(4,5)6/h9H,1-6H3

InChI key

CRHWEIDCXNDTMO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Di-tert-butylphosphine can be used in the synthesis of variety of ligands, such as Q-Phos,[1] for C-N, C-O, and C-C bond forming cross-coupling reactions.[1][2][3][4][5]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Pyr. Liq. 1 - Skin Corr. 1B

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 3

Flash Point(F)

No data available

Flash Point(C)

No data available

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A novel functionalized P,C,P pincer ligand complex.
Grimm JC, et al.
Inorganic Chemistry Communications, 3(10), 511-514 (2000)
Catalytic C-S Cross-Coupling Reactions Employing Ni Complexes of Pyrrole-Based Pincer Ligands.
Venkanna GT, et al.
ACS Catalysis, 4(9), 2941-2950 (2014)
Air stable, sterically hindered ferrocenyl dialkylphosphines for palladium-catalyzed C? C, C? N, and C? O bond-forming cross-couplings.
Kataoka N, et al.
The Journal of Organic Chemistry, 67(16), 5553-5566 (2002)
Palladium-catalyzed ?-arylation of esters and amides under more neutral conditions.
Hama T, et al.
Journal of the American Chemical Society, 125(37), 11176-11177 (2003)
Q-Phos.
William S
Aldrich Chemfiles, 7.10, 12-12 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service