Skip to Content
Merck
All Photos(1)

Documents

362654

Sigma-Aldrich

2-Fluoro-5-nitrobenzoic acid

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(NO2)CO2H
CAS Number:
Molecular Weight:
185.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

142-144 °C (lit.)

SMILES string

OC(=O)c1cc(ccc1F)[N+]([O-])=O

InChI

1S/C7H4FNO4/c8-6-2-1-4(9(12)13)3-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

ICXSHFWYCHJILC-UHFFFAOYSA-N

General description

2-Fluoro-5-nitrobenzoic acid is reported to react with an aldehyde, isonitrile and a primary amine tethered to a Boc-protected internal amino or hydroxyl nucleophile, to afford the Ugi product.

Application

2-Fluoro-5-nitrobenzoic acid may be used in the synthesis of:
  • dibenz[b,f]oxazepin-11(10H)-ones, via the nucleophilic aromatic substitution (SNAr) of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH of various 2-aminophenols on solid support
  • synthesis of substituted dibenzazocines on solid support, via SNAr of fluorine in 2-fluoro-5-nitrobenzoic acid with the OH function of the immobilized polysubstituted phenols
  • in the synthesis of oxazepines

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solid support synthesis of 2-substituted dibenz [b, f] oxazepin-11 (10H)-ones< i> via</i> S< sub> N</sub> Ar methodology on AMEBA resin.
Ouyang X, et al.
Tetrahedron, 55(10), 2827-2834 (1999)
Efficient approach for the diversity-oriented synthesis of macro-heterocycles on solid-support.
Giulianotti M and Nefzi A.
Tetrahedron Letters, 44(28), 5307-5309 (2003)
MCC/SN Ar methodology. Part 2: Novel three-step solution phase access to libraries of benzodiazepines.
Tempest P, et al.
Tetrahedron Letters, 44(9), 1947-1950 (2003)
Wim Meutermans et al.
ChemMedChem, 1(11), 1164-1194 (2006-09-20)
Drug discovery has long suffered from the difficulty of having to place pharmacophoric groups in just the right spatial arrangement to elicit the desired biological response. Although some molecule classes have been discovered that seem to be privileged structures for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service