Skip to Content
Merck
All Photos(2)

Key Documents

362069

Sigma-Aldrich

1,3-Dibenzoylpropane

98%

Synonym(s):

1,5-Diphenyl-1,5-pentanedione

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$93.20

$93.20


Estimated to ship onMay 23, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$93.20

About This Item

Linear Formula:
C6H5CO(CH2)3COC6H5
CAS Number:
Molecular Weight:
252.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$93.20


Estimated to ship onMay 23, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

66-68 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(CCCC(=O)c1ccccc1)c2ccccc2

InChI

1S/C17H16O2/c18-16(14-8-3-1-4-9-14)12-7-13-17(19)15-10-5-2-6-11-15/h1-6,8-11H,7,12-13H2

InChI key

YOLLTWVIOASMFW-UHFFFAOYSA-N

General description

1,3-Dibenzoylpropane is a 1,3-diaroylpropane. 1,3-Dibenzoylpropane is formed during hydroxocobalt(III) Schiff base complexes catalyzed selective aldol reaction of dibenzoylmethanes with formaldehyde in methanol.[1]

Application

1,3-Dibenzoylpropane may be used in the electrochemical synthesis of cis-1,2-diphenyl-1,2-cyclopentanediol, via reduction in acetonitrile.[2]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical cyclization: II. Intramolecular pinacolization of 1, 3-dibenzoylpropane in acetonitrile.
Ammar F, et al.
J. Electroanal. Chem. Interfac. Electrochem., 53(3), 407-416 (1974)
Highly selective aldol reaction of dibenzoylmethanes with formaldehyde catalyzed by cobalt Schiff base complex under neutral conditions.
Maruyama K, et al.
Tetrahedron Letters, 36(31), 5609-5612 (1995)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service