Skip to Content
Merck
All Photos(1)

Key Documents

359068

Sigma-Aldrich

(−)-Dimethyl 2,3-O-isopropylidene-L-tartrate

97%

Synonym(s):

(4R,5R)-2,2-Dimethyl-1,3-dioxolane-4,5-dicarboxylic acid dimethyl ester, Dimethyl (4R,5R)-2,2-dimethyl-1,3-dioxolane-4,5-dicarboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H14O6
CAS Number:
Molecular Weight:
218.20
Beilstein:
15406
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

optical activity

[α]20/D −54°, neat

refractive index

n20/D 1.439 (lit.)

bp

150 °C/19 mmHg (lit.)

density

1.190 g/mL at 20 °C (lit.)

functional group

ester
ether
ketal

SMILES string

COC(=O)[C@@H]1OC(C)(C)O[C@H]1C(=O)OC

InChI

1S/C9H14O6/c1-9(2)14-5(7(10)12-3)6(15-9)8(11)13-4/h5-6H,1-4H3/t5-,6-/m1/s1

InChI key

ROZOUYVVWUTPNG-PHDIDXHHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

(-)-Dimethyl 2,3-O-isopropylidene-L-tartrate may be used as a starting material in the total synthesis of (+)-migrastatin[1] and also in the synthesis of a structural analog of S-ribosylhomocysteinase (LuxS), which acts as a potent competitive inhibitor against LuxS.[2]
Valuable building block for TADDOL chiral auxiliaries,[3][4] dipyridine ligands,[5] and threitols.[6]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of substrate and intermediate analogue inhibitors of S-ribosylhomocysteinase.
Shen G, et al.
Journal of Medicinal Chemistry, 49(10), 3003-3011 (2006)
Angewandte Chemie (International Edition in English), 30, 99-99 (1991)
Organic Syntheses, 68, 92-92 (1990)
Tetrahedron Letters, 31, 7179-7179 (1990)
The total synthesis of (+)-migrastatin.
Gaul C, et al.
Journal of the American Chemical Society, 125(20), 6042-6043 (2003)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service