All Photos(1)
About This Item
Linear Formula:
C6H3(C2H5)3
CAS Number:
Molecular Weight:
162.27
Beilstein:
2040427
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
97%
refractive index
n20/D 1.501 (lit.)
bp
220-222 °C (lit.)
mp
−78 °C (lit.)
density
0.872 g/mL at 25 °C (lit.)
SMILES string
CCc1ccc(CC)c(CC)c1
InChI
1S/C12H18/c1-4-10-7-8-11(5-2)12(6-3)9-10/h7-9H,4-6H2,1-3H3
InChI key
WNLWIOJSURYFIB-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
168.8 °F - closed cup
Flash Point(C)
76 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
F Gagnaire et al.
Journal of applied toxicology : JAT, 13(2), 123-128 (1993-03-01)
1,2,4-Triethylbenzene (1,2,4-TEB) and 1,3,5-triethylbenzene (1,3,5-TEB) were administered orally to male Sprague-Dawley rats. Experimental and appropriate control rats were examined electrophysiologically for motor and sensory conduction velocities (MCV and SCV), and the amplitudes of the sensory (ASAP) and muscular action potentials
Desire D Tshala-Katumbay et al.
Acta neuropathologica, 112(3), 317-324 (2006-07-29)
The chromogenic and neurotoxic gamma-diketone 1,2-diacetylbenzene (1,2-DAB), but not its isomer 1,3-DAB, induces blue discoloration of tissues and urine, clustering of axonal microtubules and proximal neurofilament-filled axonal swellings in rodents. The remarkable chromogenic property of 1,2-DAB, a monocyclic aromatic hydrocarbon
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service