Skip to Content
Merck
All Photos(1)

Key Documents

358738

Sigma-Aldrich

Tetraoctadecylammonium bromide

98%

Synonym(s):

Tetrastearylammonium bromide

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$414.00

$414.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$414.00

About This Item

Linear Formula:
[CH3(CH2)17]4N(Br)
CAS Number:
Molecular Weight:
1107.86
Beilstein:
5846481
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$414.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

103-105 °C (lit.)

SMILES string

[Br-].CCCCCCCCCCCCCCCCCC[N+](CCCCCCCCCCCCCCCCCC)(CCCCCCCCCCCCCCCCCC)CCCCCCCCCCCCCCCCCC

InChI

1S/C72H148N.BrH/c1-5-9-13-17-21-25-29-33-37-41-45-49-53-57-61-65-69-73(70-66-62-58-54-50-46-42-38-34-30-26-22-18-14-10-6-2,71-67-63-59-55-51-47-43-39-35-31-27-23-19-15-11-7-3)72-68-64-60-56-52-48-44-40-36-32-28-24-20-16-12-8-4;/h5-72H2,1-4H3;1H/q+1;/p-1

InChI key

BUXYZRAZWWVKDS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetraoctadecylammonium bromide can be used:
  • In the synthesis of modified montmorillonite (MMT) named C72N–MMT, which can be used to prepare protein-based nanocomposite material.[1]
  • As an additive in the stabilization of liquid crystalline blue phases for optoelectronic and photonic applications.[2]
  • As a model compound in the study of the effect of quaternary ammonium salts in polymeric membranes on the functioning of ion-selective electrodes.[3] 

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The effect of lipophilic salts on surface charge in polymeric ion-selective electrodes
Zubrowska M, et al.
Electrochimica Acta, 56(17), 6114-6122 (2011)
Synthesis of nanocomposite films from wheat gluten matrix and MMT intercalated with different quaternary ammonium salts by way of hydroalcoholic solvent casting
Guilherme MR, et al.
Composites Part A: Applied Science and Manufacturing, 41(3), 375-382 (2010)
Selective stabilisation of blue phase liquid crystal induced by distinctive geometric structure of additives
Lee J, et al.
Liq. Cryst., 45(2), 230-237 (2018)

Questions

  1. Tetraoctadecylammonium bromide 98% 이 물질이 사용되는 곳이나, 용도가 무엇인지 알 수 있나요? 주로 쓰이는 곳.

    1 answer
    1. This product is used for various applications, which are typically specific to customer needs. The product description provides information on some common applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service