Skip to Content
Merck
All Photos(2)

Key Documents

358681

Sigma-Aldrich

Tetrabutylammonium p-toluenesulfonate

99%

Synonym(s):

p-Toluenesulfonic acid tetrabutylammonium salt

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$99.60
10 G
$268.00

$99.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$99.60
10 G
$268.00

About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(CH3C6H4SO3)
CAS Number:
Molecular Weight:
413.66
Beilstein:
3643278
EC Number:
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$99.60

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99%

mp

70-72 °C (lit.)

SMILES string

Cc1ccc(cc1)S([O-])(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.C7H8O3S/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;1-6-2-4-7(5-3-6)11(8,9)10/h5-16H2,1-4H3;2-5H,1H3,(H,8,9,10)/q+1;/p-1

InChI key

REAVCZWUMGIGSW-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium p-toluenesulfonate (TBAOTs) can be used as:
  • A phase transfer catalyst (PTC) in SN2 fluorinations.[1]
  • An electrolyte in the preparation of conducting polymer polypyrrole (PPy) by electrochemical polymerization technique.[2]
  • A reagent to synthesize 1-alkynyl sulfonates from 1-alkynyl-bromanes by Michael-carbene rearrangement reaction.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrabutylammonium tosylate as inert phase-transfer catalyst: The key to high efficiency SN2 radiofluorinations
Orlovskaya V, et al.
Applied Radiation and Isotopes, 109195-109195 (2020)
Actuator based on doping/undoping-induced volume change in anisotropic polypyrrole film
Okamoto T, et al.
Thin Solid Films, 393(1-2), 383-387 (2001)
Synthesis, Structure, and Reaction of 1-Alkynyl (aryl)-λ-bromanes
Ochiai M, et al.
Journal of the American Chemical Society, 125(50), 15304-15305 (2003)
Michelle L Ingalsbe et al.
Bioorganic & medicinal chemistry letters, 19(17), 4984-4987 (2009-08-08)
Due to the increasing number of strains of drug-resistant bacteria, the development of new antibiotics has become increasingly important. The antibacterial properties of quaternary amines and their derivatives on both Gram-positive and Gram-negative bacteria are well known. However, an encompassing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service