Skip to Content
Merck
All Photos(1)

Key Documents

357871

Sigma-Aldrich

5-Mercapto-1-methyltetrazole

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C2H4N4S
CAS Number:
Molecular Weight:
116.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

98%

form

powder

mp

125-128 °C (lit.)

SMILES string

Cn1nnnc1S

InChI

1S/C2H4N4S/c1-6-2(7)3-4-5-6/h1H3,(H,3,5,7)

InChI key

XOHZHMUQBFJTNH-UHFFFAOYSA-N

General description

5-Mercapto-1-methyltetrazole is a heterocyclic thiol derivative. It forms dimeric or tetrameric complexes with trimethylgallium.[1]

Application

5-Mercapto-1-methyltetrazole may be employed as heterocyclic ligand to study the geometry and stereochemical activity of the lone pair at the lead atom in hemi- and holo-directed lead(II) complexes.[2] It may be used in the chemical modification of submicron particles of mesoporous MSU-2 silica[3] and SBA-15 mesoporous silica.[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Damián Pérez-Quintanilla et al.
Journal of nanoscience and nanotechnology, 9(8), 4901-4909 (2009-11-26)
Submicron particles of mesoporous MSU-2 silica have been obtained by using Tergitol NP-9 as nonionic surfactant and TEOS as silica precursor. The material has been chemically modified with 5-mercapto-1-methyltetrazole and characterized by powder X-ray diffraction, TEM, SEM, N2 adsorption, FT-IR
K Kawamoto et al.
Japanese journal of pharmacology, 47(2), 169-178 (1988-06-01)
Liver microsomal vitamin K epoxide reductase activity was determined by measuring the formation of menaquinone-4 from the substrate menaquinone-4 2,3-epoxide. The enzyme was active when dithiothreitol (DTT) was used as a reducing agent, and the activity increased gradually with increasing
Muhammad Imran et al.
Dalton transactions (Cambridge, England : 2003), 44(3), 924-937 (2014-08-12)
To investigate the geometry and stereochemical activity of the lone pair at the lead atom, lead(ii) complexes () with one tripodal (L(1)), one dipodal (L(2)) boron-centred soft ligand and eight other small soft heterocyclic ligands, 2-mercaptobenzimidazole (L(3)), 2-mercapto-5-methylbenzimidazole (L(4)), 3-mercapto-1,2,4-triazole
J J Lipsky
Biochemical pharmacology, 38(5), 773-779 (1989-03-01)
Antibiotics that contain the 1-methyltetrazole-5-thiol (MTT) leaving group are associated with an adverse effect when alcohol is ingested after their administration. Therefore, the ability of MTT to inhibit an enzyme in alcohol metabolism, aldehyde dehydrogenase (ALDH), was examined. In the
N Kanazumi et al.
Hepato-gastroenterology, 47(36), 1695-1699 (2001-01-10)
In this study, we examined the influence of clinical treatments in the perioperative period upon PIVKA-II (plasma levels of protein induced by vitamin K absence or antagonist-II) in patients with hepatocellular carcinoma and pancreatobiliary diseases. During a perioperative period, plasma

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service