Skip to Content
Merck
All Photos(1)

Key Documents

35635

Sigma-Aldrich

cis-3,4-Dichlorocyclobutene

≥95.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4Cl2
CAS Number:
Molecular Weight:
122.98
Beilstein:
1236770
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥95.0% (GC)

refractive index

n20/D 1.503

bp

74-76 °C/55 mmHg (lit.)

density

1.302 g/mL at 20 °C (lit.)

storage temp.

2-8°C

SMILES string

Cl[C@@H]1C=C[C@@H]1Cl

InChI

1S/C4H4Cl2/c5-3-1-2-4(3)6/h1-4H/t3-,4+

InChI key

MNOAQNJDZROAPA-ZXZARUISSA-N

General description

cis-3,4-Dichlorocyclobutene is a versatile synthon in organic synthesis of natural products.[1] Infrared multiphoton excitation of cis-3,4-dichlorocyclobutene was investigated by optoacoustic detection.[2] cis-3,4-Dichlorocyclobutene undergoes 1,3-dipolar cycloaddition reaction with several nitrile oxides to yield the mixtures of syn- and anti- 4-substituted 6,7-dichloro-2-oxa-3-azabicyclo[3.2.0]hept-3-enes.[3]

Application

cis-3,4-Dichlorocyclobutene may be used in the synthesis of cyclic polyether.[1]

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

131.0 °F - closed cup

Flash Point(C)

55 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optoacoustic measurements of IR multiphoton excitation of cis-3, 4-dichlorocyclobutene.
Presser N, et al.
J. Chem. Phys. , 78(5), 2163-2169 (1983)
syn-anti-Isomerism in the cycloaddition of nitrile oxides to cis-3, 4-dichlorocyclobutene.
Bianchi G, et al.
Journal of the Chemical Society. Perkin Transactions 1, 137-140 (1974)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service