Skip to Content
Merck
All Photos(1)

Key Documents

35170

Sigma-Aldrich

2,4-Dichloroaniline

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3NH2
CAS Number:
Molecular Weight:
162.02
Beilstein:
386422
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

bp

245 °C (lit.)

mp

59-62 °C (lit.)
60-65 °C

solubility

methanol: soluble, clear, very faintly brownish-yellow

SMILES string

Nc1ccc(Cl)cc1Cl

InChI

1S/C6H5Cl2N/c7-4-1-2-6(9)5(8)3-4/h1-3H,9H2

InChI key

KQCMTOWTPBNWDB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,4-Dichloroaniline is an organic xenobiotic compound and its acute toxicity, modes of action and physicochemical properties towards the freshwater crustacean amphipod Gammarus pulex has been investigated. Biodegradation of 2,4-dichloroaniline by new strain Delftia tsuruhatensis H1 has been reported. The electrochemical oxidation of 2,4-dichloroaniline was studied in acetonitrile solution. Quantitative method for the detection of 2,4-dichloroaniline in environmental water samples by solid-phase microextraction (SPME) in gas chromatography–mass spectrometry has been reported.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The electrochemical oxidation of 4-chloroaniline, 2, 4-dichloroaniline and 2, 4, 6-trichloroaniline in acetonitrile solution.
Kadar M, et al.
Electrochimica Acta, 46(9), 1297-1306 (2001)
Determination of aromatic amines by solid-phase microextraction and gas chromatography−mass spectrometry in water samples.
Muller L, et al.
Journal of Chromatography A, 791(1), 221-230 (1997)
Phototransformation of 2,4-dichloroaniline in a surface freshwater environment: effects on microbial assemblages.
H M Hwang et al.
Bulletin of environmental contamination and toxicology, 60(1), 81-87 (1998-03-04)
Sophie Pascal-Lorber et al.
Journal of agricultural and food chemistry, 51(16), 4712-4718 (2004-01-07)
The uptake and metabolism of [14C]-2,4-dichlorophenol (DCP) and [14C]-2,4-dichloroaniline (DCA) were investigated in wheat and soybean. Seeds were exposed to a nutrient solution containing 50 microM of one of two radiolabeled compounds, and plant organs were harvested separately after 18
Michael Kilemade et al.
Aquatic toxicology (Amsterdam, Netherlands), 63(3), 207-219 (2003-04-25)
Interest in and concern for the quality of the environment has prompted a great deal of research into methods of measuring and assessing changes in it. One problem of major interest is that of increasing amounts of mutagenic/carcinogenic chemicals generated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service