Skip to Content
Merck
All Photos(1)

Key Documents

348341

Sigma-Aldrich

Cerium(IV) oxide

powder, 90%

Synonym(s):

Ceric oxide, Ceric oxide, ceria

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CeO2
CAS Number:
Molecular Weight:
172.11
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

90%

form

powder

reaction suitability

reagent type: catalyst
core: cerium

avg. part. size

3 μm

density

7.13 g/mL at 25 °C (lit.)

SMILES string

O=[Ce]=O

InChI

1S/Ce.2O

InChI key

CETPSERCERDGAM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

The additon of CeO2 to CuO/TiO2 increases specific surface area and inhibits sintering and pore size growth at high temperatures.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lijuan Zhao et al.
ACS nano, 6(11), 9615-9622 (2012-10-12)
The rapid development of nanotechnology will inevitably release nanoparticles (NPs) into the environment with unidentified consequences. In addition, the potential toxicity of CeO(2) NPs to plants and the possible transfer into the food chain are still unknown. Corn plants (Zea
Hailong Li et al.
Journal of hazardous materials, 243, 117-123 (2012-11-08)
MnO(x)-CeO(2) mixed-oxide supported on TiO(2) (Mn-Ce/Ti) was synthesized by an ultrasound-assisted impregnation method and employed to oxidize elemental mercury (Hg(0)) at 200°C in simulated coal combustion flue gas. Over 90% of Hg(0) oxidation was achieved on the Mn-Ce/Ti catalyst at
Ashraf Aly Hassan et al.
Journal of hazardous materials, 244-245, 251-258 (2012-12-29)
Experimental and computational investigation of the transport parameters of nanoparticles (NPs) flowing through porous media has been made. This work intends to develop a simulation applicable to the transport and retention of NPs in saturated porous media for investigating the
Peng Zhang et al.
ACS nano, 6(11), 9943-9950 (2012-10-27)
Biotransformation is a critical factor that may modify the toxicity, behavior, and fate of engineered nanoparticles in the environment. CeO(2) nanoparticles (NPs) are generally recognized as stable under environmental and biological conditions. The present study aims to investigate the biotransformation
Dengsong Zhang et al.
Nanoscale, 5(3), 1127-1136 (2013-01-04)
The MnO(x) and CeO(x) were in situ supported on carbon nanotubes (CNTs) by a poly(sodium 4-styrenesulfonate) assisted reflux route for the low-temperature selective catalytic reduction (SCR) of NO with NH(3). X-Ray diffraction (XRD), transmission electron microscopy (TEM), high-resolution TEM (HRTEM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service