Skip to Content
Merck
All Photos(2)

Documents

348295

Sigma-Aldrich

4-Morpholinecarbonyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8ClNO2
CAS Number:
Molecular Weight:
149.58
Beilstein:
116257
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

liquid

refractive index

n20/D 1.498 (lit.)

bp

137-138 °C/33 mmHg (lit.)

density

1.282 g/mL at 25 °C (lit.)

SMILES string

ClC(=O)N1CCOCC1

InChI

1S/C5H8ClNO2/c6-5(8)7-1-3-9-4-2-7/h1-4H2

InChI key

XMWFMEYDRNJSOO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Morpholinecarbonyl chloride is a photographic intermediate. Rates of solvolysis of 4-morpholinecarbonyl chloride with N-alkyl group in aqueous binary solvents of acetone, ethanol, methanol, methanol-d, 2,2,2-trifluoroethanol, pure water and D2O has been determined. Preparation of 4-morpholinecarbonyl chloride has been reported.

Application

4-Morpholinecarbonyl chloride may be used in the preparation of a versatile fluorogenic label for biomolecular imaging. It may be used in the synthesis of:
  • morpholine-4-carboxylic acid [4-(2,4-dimethylphenyl)thiazol-2-yl]amide
  • morpholine-4-carboxylic acid [4-(2,4,6-trimethylphenyl)thiazol-2-yl]amide
  • 2-isopropyl-6-methylpyrimidin-4-yl morpholine-4-carboxylate

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Skin Irrit. 2

Supplementary Hazards

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Correlation of the rates of solvolysis of 4-morpholinecarbonyl chloride using the extended Grunwald-Winstein equation.
Kim R, et al.
Bull. Korean Chem. Soc., 31(7), 1963-1963 (2010)
Synthesis and antifungal activities of some novel pyrimidine derivatives.
Sun L, et al.
Molecules (Basel), 16(7), 5618-5628 (2011)
Xiao-Long Qiu et al.
Journal of medicinal chemistry, 52(6), 1757-1767 (2009-02-27)
High expression in cancer 1 (Hec1) is an oncogene overly expressed in many human cancers. Small molecule inhibitor of Nek2/Hec1 (INH) targeting the Hec1 and its regulator, Nek2, in the mitotic pathway, was identified to inactivate Hec1/Nek2 function mediated by
Luke D Lavis et al.
ACS chemical biology, 1(4), 252-260 (2006-12-14)
Traditional small-molecule fluorophores are always fluorescent. This attribute can obscure valuable information in biological experiments. Here, we report on a versatile "latent" fluorophore that overcomes this limitation. At the core of the latent fluorophore is a derivative of rhodamine in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service