Skip to Content
Merck
All Photos(1)

Key Documents

348074

Sigma-Aldrich

2-Chloro-4-fluorobenzaldehyde

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$55.20

$55.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$55.20

About This Item

Linear Formula:
ClC6H3(F)CHO
CAS Number:
Molecular Weight:
158.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$55.20

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

solid

bp

118-120 °C/50 mmHg (lit.)

mp

60-63 °C (lit.)

functional group

aldehyde
chloro
fluoro

SMILES string

Fc1ccc(C=O)c(Cl)c1

InChI

1S/C7H4ClFO/c8-7-3-6(9)2-1-5(7)4-10/h1-4H

InChI key

KMQWNQKESAHDKD-UHFFFAOYSA-N

General description

2-Chloro-4-fluorobenzaldehyde is a halogen substituted benzaldehyde.

Application

2-Chloro-4-fluorobenzaldehyde may be used in the synthesis of substituted α-cyanocinnamic acid, via Knoevenagel condensation reaction.[1] It may be used in the synthesis of 2-(2-benzimidazolyl)-3-(2-chloro-4-fluorophenyl)acrylonitrile.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biological activity and DNA binding studies of 2-substituted benzimidazo [1, 2-a] quinolines bearing different amino side chains.
Perin N, et al.
MedChemComm, 4(12), 1537-1550 (2013)
Thorsten W Jaskolla et al.
Proceedings of the National Academy of Sciences of the United States of America, 105(34), 12200-12205 (2008-08-30)
Matrix-assisted laser desorption ionization (MALDI) has become an enabling technology for the fields of protein mass spectrometry (MS) and proteomics. Despite its widespread use, for example, in protein identification via peptide mass fingerprinting, a comprehensive model for the generation of

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service