Skip to Content
Merck
All Photos(1)

Key Documents

345482

Sigma-Aldrich

Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate

98%

Synonym(s):

α,β-Isopropylidene-D-glyceric acid methyl ester, Methyl α,β-isopropylidene-D-glycerate, Methyl 2,3-O-isopropylidene-D-glycerate

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$168.00

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$168.00

About This Item

Empirical Formula (Hill Notation):
C7H12O4
CAS Number:
Molecular Weight:
160.17
Beilstein:
4292353
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$168.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

optical activity

[α]20/D +8.5°, c = 1.5 in acetone

refractive index

n20/D 1.426 (lit.)

bp

70-75 °C/10 mmHg (lit.)

density

1.106 g/mL at 25 °C (lit.)

functional group

ester
ether
ketal

SMILES string

COC(=O)[C@H]1COC(C)(C)O1

InChI

1S/C7H12O4/c1-7(2)10-4-5(11-7)6(8)9-3/h5H,4H2,1-3H3/t5-/m1/s1

InChI key

DOWWCCDWPKGNGX-RXMQYKEDSA-N

Application

Methyl (R)-(+)-2,2-dimethyl-1,3-dioxolane-4-carboxylate may be used as a starting material to synthesize (2R)-[1-2H2]-glycerol, a probe to study the stereochemistry of glycerol metabolism in Streptomyces cattleya.[1] It may also be used to synthesize organic building blocks like (R)-2,3-dihydroxy-3-methylbutyl toluene-p-sulfonate[2] and (2S,8R)-2-amino-8-[(R) 2,2-dimethyl-1,3-dioxolan-4-yl]-8-oxooctanoic acid.[3]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Biosynthesis of fluoroacetate and 4-fluorothreonine by Streptomyces cattleya. The stereochemical processing of glycerol.
Nieschalk J, et al.
Chemical Communications (Cambridge, England), 8, 799-800 (1997)
Synthesis of 2-amino-8-oxodecanoic acids (Aodas) present in natural hystone deacetylase inhibitors.
Rodriquez M, et al.
The Journal of Organic Chemistry, 71(1) (2006)
Synthesis of (24 R)-11a-(4-carboxybutyryloxy)-24, 25-dihydroxyvitamin D 3: a novel haptenic derivative producing antibodies of high affinity for (24 R)-24, 25-dihydroxyvitamin D 3.
Kobayashi N, et al.
J. Chem. Soc., Perkin Trans., 3, 269-275 (1994)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service