Skip to Content
Merck
All Photos(2)

Key Documents

345164

Sigma-Aldrich

Tin(II) acetate

Synonym(s):

Stannous acetate, Tin acetate, Tin diacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Sn(CH3CO2)2
CAS Number:
Molecular Weight:
236.80
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

form

solid

Quality Level

reaction suitability

core: tin
reagent type: catalyst

mp

180-182 °C (lit.)

SMILES string

CC(=O)O[SnH2]OC(C)=O

InChI

1S/2C2H4O2.Sn/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

PNOXNTGLSKTMQO-UHFFFAOYSA-L

General description

Tin(II) acetate can be used as a photocatalyst to produce Arylstannane(IV) Reagents.[1]

Application

  • Enhances the rate of thermal depolymerization of poly(lactic acid) fibers
  • Reactant for the synthesis of Sn-Cu bimetallic nanoparticles
  • used in in preparation of tin anode by organic electroplating for rechargeable thin-film batteries
  • Used as tin source for preparation of high surface area tin oxide catalysts
Used to prepare tin(IV) oxide thin films by photochemical vapor deposition.[2] With palladium(II) acetate, an efficient catalytic system for C-H activation of the methoxy group of anisole.[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junshan Li et al.
ChemSusChem, 12(7), 1451-1458 (2019-01-25)
Co-Sn solid-solution nanoparticles with Sn crystal structure and tuned metal ratios were synthesized by a facile one pot solution-based procedure involving the initial reduction of a Sn precursor followed by incorporation of Co within the Sn lattice. These nanoparticles were
Gianvito Caputo et al.
Chemphyschem : a European journal of chemical physics and physical chemistry, 18(12), 1635-1641 (2017-04-04)
The localized in situ formation of tin dioxide (SnO
The Journal of Organic Chemistry, 59, 7521-7521 (1994)
Photocatalytic Reaction of Aryl Halides with Tin (II) Acetate to Generate Arylstannane (IV) Reagents
Zemtsov AA, et al.
ACS Catalysis, 13, 12766-12773 (2023)
The Journal of Organic Chemistry, 59, 7521-7521 (1994)

Questions

1–3 of 3 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  3. what is the purity percentage?

    1 answer
    1. The purity of this product is not determined. The Tin(II) content is tested on lot-specific bases with a specification of 49 - 53%.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service