Skip to Content
Merck
All Photos(1)

Key Documents

344281

Sigma-Aldrich

2,4,6-Trichlorobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl3C6H2CO2H
CAS Number:
Molecular Weight:
225.46
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

mp

160-164 °C (lit.)

functional group

carboxylic acid
chloro

SMILES string

OC(=O)c1c(Cl)cc(Cl)cc1Cl

InChI

1S/C7H3Cl3O2/c8-3-1-4(9)6(7(11)12)5(10)2-3/h1-2H,(H,11,12)

InChI key

RAFFVQBMVYYTQS-UHFFFAOYSA-N

General description

Structure and hydrogen bonding pattern in 2,4,6-trichlorobenzoic acid is reported.[1]

Application

2,4,6-Trichlorobenzoic acid may be employed as sole carbon and energy supplement for a microbial community.[2] It may be used in the synthesis of (+)-methynolide, the aglycon of a macrolide antibiotic, methymycin.[3]
Reactant involved in:
  • Active-sodium-promoted reductive cleavage of halogenated benzoic acids
  • Synthesis of aryl aminopyrazole benzamides for use as non-steroidal selective glucocorticoid receptor agonists
  • Flame retardant monomer synthesis
  • Synthesis of 3,4,7-trisubstituted coumarins for use as antifungals
  • Solid-phase synthesis of saphenamycin analogs with antimicrobial activity

Cocatalyst for cis-dihydroxylation and epoxidation of alkenes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F R Johannsen et al.
Journal of applied toxicology : JAT, 7(1), 67-70 (1987-02-01)
The acute rat oral LD50 of 2,4,6-Trichlorobenzyl chloride (TCBC) was determined to be 3075 mg/kg. When male and female rats were administered 1500 and 3000 ppm TCBC in the diet for 3 weeks, marked retardation in weight gain was observed.
S Moller et al.
Applied and environmental microbiology, 63(6), 2432-2438 (1997-06-01)
A microbial community was cultivated in flow cells with 2,4,6-trichlorobenzoic acid (2,4,6-TCB) as sole carbon and energy source and was examined with scanning confocal laser microscopy and fluorescent molecular probes. The biofilm community which developed under these conditions exhibited a
2, 4, 6-Trichlorobenzoic acid: Structure and hydrogen-bonding pattern.
Lalancette RA, et al.
Acta Crystallographica Section C, Structural Chemistry, 52(7), 1801-1804 (1996)
Hong-Se Oh et al.
Organic & biomolecular chemistry, 7(21), 4458-4463 (2009-10-16)
Methynolide and 10-epi-methynolide were synthesized from the necessary segments, which were prepared by the addition of Grignard reagents to the corresponding alpha-alkoxyketones utilizing 1,2-stereochemical selection based on Cram chelation control. Ring-closing metathesis, as the key reaction, was carried out to
Kevin X Zhang et al.
Nature, 585(7825), 420-425 (2020-09-04)
The opsin family of G-protein-coupled receptors are used as light detectors in animals. Opsin 5 (also known as neuropsin or OPN5) is a highly conserved opsin that is sensitive to visible violet light1,2. In mice, OPN5 is a known photoreceptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service