Skip to Content
Merck
All Photos(1)

Key Documents

342785

Sigma-Aldrich

Magnesium oxide

fused, pieces, 3-12 mm, 99.95% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.95% trace metals basis

form

fused
pieces

particle size

3-12 mm

mp

2852 °C (lit.)

application(s)

battery manufacturing

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for vacuum deposition.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

  • Choose from one of the most recent versions:

    Certificates of Analysis (COA)

    Lot/Batch Number

    Don't see the Right Version?

    If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Morshed Khandaker et al.
    Journal of biomechanics, 46(5), 1035-1039 (2013-01-22)
    The objective of this study was to determine whether inclusion of magnesium oxide (MgO) in micro and nanoparticulate forms in poly methyl methacrylate (PMMA) cement has any influence on the fracture toughness of bone-cement interfaces. An interfacial fracture mechanics technique
    Dalia Latkauskiene et al.
    Progress in orthodontics, 13(2), 100-108 (2012-10-02)
    To evaluate the clinical performance of the stainless steel crown Herbst (cHerbst) used as a single appliance for a single phase therapy. A total of 180 consecutive Class II patients were treated with cHerbst for one year and followed up
    Sanket A Deshmukh et al.
    Physical chemistry chemical physics : PCCP, 14(44), 15593-15605 (2012-10-19)
    Atomic scale characterization of the structure and dynamics of confined water molecules located near the metal oxide-aqueous interface is carried out using molecular dynamics simulations. Proximity effects on water molecules (H(2)O) near a magnesium oxide surface (MgO(100)) at room temperature
    Jian-Bin Shi et al.
    Carbohydrate research, 359, 65-69 (2012-08-29)
    This work describes the structural changes of bagasse hemicelluloses during the cooking process involving active oxygen (O(2) and H(2)O(2)) and solid alkali (MgO). The hemicelluloses obtained from the bagasse raw material, pulp, and yellow liquor were analyzed by high-performance anion-exchange
    C B Abi et al.
    Journal of the mechanical behavior of biomedical materials, 18, 123-131 (2012-12-25)
    The aim of the present study was to investigate the production of tetragonal zirconia (t-ZrO(2)) particles (experimental t-ZrO(2)) from monoclinic zirconia (m-ZrO(2)) and to evaluate the effect of the t-ZrO(2) content on the fracture toughness of alumina-zirconia composites by conducting

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service