Skip to Content
Merck
All Photos(2)

Documents

342440

Sigma-Aldrich

Cobalt(II) hydroxide

technical grade, 95%

Synonym(s):

Cobalt dihydroxide, Cobalt(2+) hydroxide, Cobaltous hydroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(OH)2
CAS Number:
Molecular Weight:
92.95
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Assay

95%

form

powder

density

3.597 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

SMILES string

O[Co]O

InChI

1S/Co.2H2O/h;2*1H2/q+2;;/p-2

InChI key

ASKVAEGIVYSGNY-UHFFFAOYSA-L

Related Categories

General description

Cobalt(II) hydroxide is a versatile compound with various applications in battery manufacturing, sensors, supercapacitor, and magnetic materials. It is also used as a key component in the production of rechargeable lithium-ion batteries. Additionally, it is also used in electrochemical reactions, including the oxidation of glucose and the oxygen evolution reaction (OER).

Application

Cobalt(II) hydroxide can be used in the research and development :
  • As an additive in Ni-based rechargeable alkaline batteries, including Ni-Cd and Ni-MH batteries, to improve their electrochemical performance.   
  • To synthesize reduced graphene oxide/cobalt hydroxide (rGO/Co(OH)2) composite material, which exhibits enhanced electrochemical performance for supercapacitor applications.     
  • As a electrode material for high-performance supercapacitors. It exhibits high specific capacitance and long cycling stability, making it a promising electrode material for supercapacitors.

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Carc. 1B - Eye Irrit. 2 - Repr. 1B - Resp. Sens. 1B - Skin Sens. 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claudia Backes et al.
ACS nano, 13(6), 7050-7061 (2019-06-15)
Liquid phase exfoliation is a commonly used method to produce 2D nanosheets from a range of layered crystals. However, such nanosheets display broad size and thickness distributions and correlations between area and thickness, issues that limit nanosheet application potential. To

Articles

Lithium-ion batteries' characteristics make them popular for electricity storage due to portability, rechargeability, and low cost.

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service