Skip to Content
Merck
All Photos(1)

Key Documents

342157

Sigma-Aldrich

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde

99%

Synonym(s):

5-(Trifluoromethoxy)salicylaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H3(OH)CHO
CAS Number:
Molecular Weight:
206.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

82 °C/60 mmHg (lit.)

mp

31-33 °C (lit.)

SMILES string

Oc1ccc(OC(F)(F)F)cc1C=O

InChI

1S/C8H5F3O3/c9-8(10,11)14-6-1-2-7(13)5(3-6)4-12/h1-4,13H

InChI key

WQUZBERVMUEJTD-UHFFFAOYSA-N

General description

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde is formed as intermediate during the biotransformation pathways of CP-122,721 in humans.[1]

Application

2-Hydroxy-5-(trifluoromethoxy)benzaldehyde may be used in the preparation of :
  • 2-[(E)-2-hydroxy-5-(trifluoromethoxy)benzylideneamino]-4-methylphenol[2]
  • (E)-2-((3-fluorophenylimino)methyl)-4-(trifluoromethoxy) phenol[3]
  • 2-[(E)-(naphthalen-2-ylimino) methyl]-4-(trifluoromethoxy) phenol[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Aslı Tosyalı Karadağ et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 1), o95-o95 (2010-01-01)
The title compound, C(15)H(12)F(3)NO(3), is a Schiff base which adopts the cis-quinoid form in the solid state. The dihedral angle between the least-squares planes of the benzene rings being 3.6 (1)°. The F atoms of the -CF(3) group are disordered over
Yelda Bingöl Alpaslan et al.
Acta crystallographica. Section E, Structure reports online, 66(Pt 3), o510-o510 (2010-01-01)
The title compound, C(14)H(9)F(4)NO(2), is a Schiff base which adopts the phenol-imine tautomeric form in the solid state. The H atom is located on the hydr-oxy O atom rather than on the N atom. This H atom is involved in
Kevin Colizza et al.
Drug metabolism and disposition: the biological fate of chemicals, 35(6), 884-897 (2007-03-16)
The metabolism, pharmacokinetics, and excretion of a potent and selective substance P receptor antagonist, CP-122,721 [(+)-(2S,3S)-3-(2-methoxy-5-trifluoromethoxybenzylamino)-2-phenylpiperidine], have been studied in six healthy male human subjects [four extensive metabolizers (EMs) and two poor metabolizers (PMs) of CYP2D6) following oral administration of
2-[(E)-(Naphthalen-2-ylimino) methyl]-4-(trifluoromethoxy) phenol.
Pekdemir M, et al.
Acta Crystallographica Section E, Structure Reports Online, 68(4), o10204-o10204 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service