Skip to Content
Merck
All Photos(1)

Key Documents

341096

Sigma-Aldrich

Succinimidyl 2,2,2-trichloroethyl carbonate

98%

Synonym(s):

N-(2,2,2-Trichloroethoxycarbonyloxy)succinimide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
133,30 €
1 G
182,80 €
2 G
314,80 €
5 G
644,80 €
10 G
1106,80 €
20 G
2030,80 €
25 G
2426,80 €

Vendor SKU: NC-0225-500MG

133,30 €


Ships in 21 days - from Aldrich Partner.


Select a Size

Change View
500 MG
133,30 €
1 G
182,80 €
2 G
314,80 €
5 G
644,80 €
10 G
1106,80 €
20 G
2030,80 €
25 G
2426,80 €

About This Item

Empirical Formula (Hill Notation):
C7H6Cl3NO5
CAS Number:
Molecular Weight:
290.49
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Vendor SKU: NC-0225-500MG

133,30 €


Ships in 21 days - from Aldrich Partner.

Assay

98%

mp

111-113 °C (lit.)

storage temp.

2-8°C

SMILES string

ClC(Cl)(Cl)COC(=O)ON1C(=O)CCC1=O

InChI

1S/C7H6Cl3NO5/c8-7(9,10)3-15-6(14)16-11-4(12)1-2-5(11)13/h1-3H2

InChI key

WBZXNGAFYBGQFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Succinimidyl 2,2,2-trichloroethyl carbonate was used in the synthesis of:
  • methyl [phenyl 5-(2,2,2-trichloroethoxycarbonylamino)-3,5-dideoxy-2-thio-β-D-glycero-D-galacto-2-nonulopyranosid]-onate[1]
  • phenyl 3,4-di-O-benzyl-6-O-(tert-butyl-dimethyl-silyl)-1-selanyl-2-(2,2,2-trichloroethoxycarbonylamine)-β-D-galactopyranoside[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chien-Tai Ren et al.
The Journal of organic chemistry, 72(14), 5427-5430 (2007-06-21)
Synthesis of a tetrasaccharide glycosyl glycerol, the core structure of glycoglycerolipid from Meiothermus taiwanensis ATCC BAA-400, was described. A one-pot glycosylation with three components was employed as a key step.
Chien-Tai Ren et al.
The Journal of organic chemistry, 67(4), 1376-1379 (2002-02-16)
The preparation of a disaccharide 2, Neu5Ac-alpha-(2-->5)Neu5Gc having a alpha-benzyl protecting group at the reducing end, by the coupling of the easily accessible building units 4 and 5 is described. Subsequent deprotection of the coupling adduct led to the isolation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service