Skip to Content
Merck
All Photos(1)

Key Documents

340480

Sigma-Aldrich

trans,trans-Farnesyl acetate

95%

Synonym(s):

(E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-ol acetate, (E,E)-3,7,11-Trimethyl-2,6,10-dodecatrien-1-yl acetate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$284.00

$284.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Select a Size

Change View
10 G
$284.00

About This Item

Linear Formula:
CH3CO2CH2CH=C(CH3)CH2CH2CH=C(CH3)CH2CH2CH=C(CH3)2
CAS Number:
Molecular Weight:
264.40
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$284.00


Estimated to ship onApril 10, 2025


Request a Bulk Order

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.477 (lit.)

bp

115-125 °C/0.3 mmHg (lit.)

density

0.914 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CC(=O)OC\C=C(/C)CC\C=C(/C)CC\C=C(\C)C

InChI

1S/C17H28O2/c1-14(2)8-6-9-15(3)10-7-11-16(4)12-13-19-17(5)18/h8,10,12H,6-7,9,11,13H2,1-5H3/b15-10+,16-12+

InChI key

ZGIGZINMAOQWLX-NCZFFCEISA-N

General description

trans,trans-Farnesyl acetate is one of the most odor-active or key compounds of Hyuganatsu aroma in Hyuganatsu (Citrus tamurana Hort. ex Tanaka) peel oil.[1] Asymmetric dihydroxylation of trans,trans-farnesyl acetate using the new Sharpless′ ligand has been studied.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Asymmetric dihydroxylation of geranyl, neryl and< i> trans, trans</i>-farnesyl acetates.
Vidari G, et al.
Tetrahedron Letters, 34(40), 6485-6488 (1993)
H S Choi et al.
Journal of agricultural and food chemistry, 49(5), 2404-2408 (2001-05-23)
The volatile components of Hyuganatsu (Citrus tamurana Hort. ex Tanaka) peel oil, isolated by cold-pressing, were investigated by chemical and sensory analyses. According to chemical analysis by GC and GC-MS, limonene (84.0%) was the most abundant compound, followed by gamma-terpinene
Pingping Liu et al.
Pest management science, 76(9), 2986-2993 (2020-04-05)
Thrips, Megalurothrips usitatus, usually display aggregation behavior, which is probably mediated by a male-produced aggregation pheromone. Aggregation pheromones are species-specific, and can be used to develop commercial lures for monitoring and mass-trapping of pests. The active components of the aggregation
Fangfang Zeng et al.
Insect biochemistry and molecular biology, 113, 103213-103213 (2019-08-24)
Mosquitoes rely heavily on the olfactory system to find a host for a bloodmeal, plants for a source of energy and suitable sites for oviposition. Here, we examined a cluster of eight odorant receptors (ORs), which includes one OR, CquiOR1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service