Skip to Content
Merck
All Photos(3)

Documents

340073

Sigma-Aldrich

Ethylenediaminetetraacetic acid calcium disodium salt hydrate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ca(O2CCH2)2NCH2CH2N(CH2CO2Na)2·xH2O
CAS Number:
Molecular Weight:
374.27 (anhydrous basis)
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

SMILES string

O.[Na]OC(=O)CN(CCN1CC(=O)O[Ca]OC(=O)C1)CC(=O)O[Na]

InChI

1S/C10H16N2O8.Ca.2Na.H2O/c13-7(14)3-11(4-8(15)16)1-2-12(5-9(17)18)6-10(19)20;;;;/h1-6H2,(H,13,14)(H,15,16)(H,17,18)(H,19,20);;;;1H2/q;+2;2*+1;/p-4

InChI key

JHECKPXUCKQCSH-UHFFFAOYSA-J

Application

Ethylenediaminetetraacetic acid calcium disodium salt hydrate is calcium and sodium salt of EDTA and it can be used:
  • To retain the quality of litchi fruit by the postharvest treatment process.
  • In the preparation of an aqueous solution for hydroxyapatite coating, which is used to coat the magnesium material/particle for biomedical applications.
  • To prepare a mobile phase for the separation of diol products from the aqueous solution of the reaction mixture by chromatography techniques.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Influence of modified atmosphere packaging and postharvest treatments on quality retention of litchi cv. Mauritius
Sivakumar D and Korsten L
Postharvest biology and technology, 41(2), 135-142 (2006)
Production and bio-corrosion resistance of porous magnesium with hydroxyapatite coating for biomedical applications
Kang M, et al.
Materials Letters, 108, 122-124 (2013)
Direct conversion of carbohydrates to diol by the combination of niobic acid and a hydrophobic ruthenium catalyst
Duan Y, et al.
Royal Society of Chemistry Advances, 7(42), 26487-26493 (2017)
Polyetheretherketone/magnesium composite selectively coated with hydroxyapatite for enhanced in vitro bio-corrosion resistance and biocompatibility
Jung H, et al.
Materials Letters, 116, 20-22 (2014)
Temmei Ito et al.
Biomedical materials (Bristol, England), 10(1), 015025-015025 (2015-03-03)
In order to obtain a hydroxyapatite (HAp)-poly(ethylene glycol) (PEG) composite, tetra amine-terminated PEG was crosslinked using disuccinimidyl tartrate to obtain a PEG hydrogel. Using two kinds of chelators with different stability constants for Ca ion (N-(2-hydroxyethyl) ethylenediamine-N,N',N'-triacetic acid (EDTA-OH, 8.14)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service