Skip to Content
Merck
All Photos(1)

Key Documents

339938

Sigma-Aldrich

3,4-Diaminotoluene

99%, purified by sublimation

Synonym(s):

4-Methyl-o-phenylenediamine, 3,4-Toluenediamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NH2)2
CAS Number:
Molecular Weight:
122.17
Beilstein:
507965
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

purified by

sublimation

bp

155-156 °C/18 mmHg (lit.)

mp

87-89 °C (lit.)

SMILES string

Cc1ccc(N)c(N)c1

InChI

1S/C7H10N2/c1-5-2-3-6(8)7(9)4-5/h2-4H,8-9H2,1H3

InChI key

DGRGLKZMKWPMOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Diaminotoluene on treatment with [{PhP(Se)(μ-Se)}2], causes the rupture of the dimer, giving PhP(Se)(NHC6H3(CH3-3)NH-1,2).

Application

3,4-Diaminotoluene was employed as catalyst for the two-step reduction of Zn(II) ions at dropping mercury electrode in 1M NaClO4 + 0.001M HClO4. It was also used in the synthesis of new benzimidazole derivatives with cyclohexylalkyl and aralkyl substituents in position 2.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bridge cleavage of [{PhP(Se)(?-Se)}2] by 1,2- C6H4(EH)(E'H) (E, E'=O or NH). X-ray crystal structure of PhP(Se)(NHC6H4NH-1,2).
Bhattacharyya P, et al.
Journal of Organometallic Chemistry, 623(1), 116-119 (2001)
The catalysis of the reduction of Zn (II) ions by 3, 4-diaminotoluene.
Dalmata G.
Electrochimica Acta, 42(9), 1307-1314 (1997)
Synthesis and tuberculostatic activity of new benzimidazole derivatives.
Foks H, et al.
Chemistry of Heterocyclic Compounds, 42(5), 611-614 (2006)
Effect of 4 toluene diamine isomers on murine testicular DNA synthesis.
E J Greene et al.
Mutation research, 91(1), 75-79 (1981-01-01)
T A Marks et al.
Teratology, 24(3), 253-265 (1981-12-01)
Pregnant outbred albino (CD-1) mice were given 2-nitro-p-phenylenediamine (2NPPD; 32-256 mg/kg/day), 4-nitro-o-phenylenediamine (4NOPD; 16-1024 mg/kg/day) or 2,5-toluenediamine sulfate (2,5TDS; 16-64 mg/kg/day) by subcutaneous injection on Days 6-15 of gestation. The mice were killed on Day 18, the general health and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service