Skip to Content
Merck
All Photos(1)

Key Documents

339563

Sigma-Aldrich

2,3,4,5-Tetrafluorobenzoyl chloride

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$44.10

$44.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$44.10

About This Item

Linear Formula:
HC6F4COCl
CAS Number:
Molecular Weight:
212.53
Beilstein:
5266860
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$44.10

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.4787 (lit.)

density

1.58 g/mL at 25 °C (lit.)

functional group

acyl chloride
fluoro

SMILES string

Fc1cc(C(Cl)=O)c(F)c(F)c1F

InChI

1S/C7HClF4O/c8-7(13)2-1-3(9)5(11)6(12)4(2)10/h1H

InChI key

XWCKIXLTBNGIHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2,3,4,5-Tetrafluorobenzoyl chloride was used in the preparation of 4-hydroxy-1-(3-pyridyl)-1-butanone-tetrafluorobenzoate.[1] It was also used in the synthesis of N-[(4-carbamoylphenyl)carbamothioyl]-2,3,4,5-tetrafluorobenzamide.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S G Carmella et al.
Cancer research, 50(17), 5438-5445 (1990-09-01)
Hemoglobin adducts of the carcinogenic tobacco-specific nitrosamines 4-(methylnitrosamino)-1(3-pyridyl)-1-butanone and N'-nitrosonornicotine were quantified in blood samples collected from snuff dippers, smokers, and nonsmokers. Mild base treatment of hemoglobin adducted by 4-(methylnitrosamino)-1-(3-pyridyl)-1-butanone or N'-nitrosonornicotine releases 4-hydroxy-1-(3-pyridyl)-1-butanone (HPB). HPB was enriched by solvent
Li-Dan Zhang et al.
Acta crystallographica. Section E, Structure reports online, 67(Pt 3), o688-o688 (2011-04-28)
In the title compound, C(15)H(9)F(4)N(3)O(2)S, the N,N'-disubstituted thio-urea fragment adopts a cis,trans geometry, stabilized by an intra-molecular N-H⋯O hydrogen bond to the carbonyl O atom of the tetra-fluoro-benzoyl group. The central thio-urea group makes dihedral angles of 47.79 (7) and 35.54 (8)°

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service