Skip to Content
Merck
All Photos(1)

Key Documents

337021

Sigma-Aldrich

2-Bromo-3-methylthiophene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5BrS
CAS Number:
Molecular Weight:
177.06
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.572 (lit.)

bp

173-176 °C (lit.)

density

1.572 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

Cc1ccsc1Br

InChI

1S/C5H5BrS/c1-4-2-3-7-5(4)6/h2-3H,1H3

InChI key

YYJBWYBULYUKMR-UHFFFAOYSA-N

General description

2-Bromo-3-methylthiophene is formed (approx. 90%) by bromination of 3-methyl thiophene with N-bromosuccinimide in the absence of benzoyl peroxide.[1]

Application

2-Bromo-3-methylthiophene was used in the preparation of poly(3,3′′′-dimethyl-(2,2′:5′,2′:5′,2′′′)tetrathiophene)).[2] It was also used in the preparation of 2-bromo-3-(bromomethyl)thiophene, lachrymatory compound.[3]

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Regioregular poly (thiophene-3-alkanoic acid) s: water soluble conducting polymers suitable for chromatic chemosensing in solution and solid state.
Ewbank PC, et al.
Tetrahedron, 60(49), 11269-11275 (2004)
The Effect of Benzoyl Peroxide on the Bromination of Methylthiophenes by N-Bromosuccinimide
Dittmer K, et al.
Journal of the American Chemical Society, 71(4), 1201-1204 (1949)
Structural studies of poly (dimethyl-tetrathiophene, an intermediate between poly (thiophene) and poly (3-methylthiophene).
Fell HJ, et al.
Synthetic Metals, 55(1), 420-425 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service