Skip to Content
Merck
All Photos(2)

Key Documents

336513

Sigma-Aldrich

(S)-(+)-1-Cyclohexylethylamine

98%

Synonym(s):

(S)-(+)-α-Methylcyclohexanemethylamine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$121.00

$121.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$121.00

About This Item

Linear Formula:
C6H11CH(CH3)NH2
CAS Number:
Molecular Weight:
127.23
Beilstein:
2935069
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.22

$121.00


Estimated to ship onMay 22, 2025


Request a Bulk Order

Assay

98%

form

liquid

optical activity

[α]25/D +3.5°, neat

optical purity

ee: 95% (GLC)

refractive index

n20/D 1.4614 (lit.)

bp

60 °C/12 mmHg (lit.)

density

0.856 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

C[C@H](N)C1CCCCC1

InChI

1S/C8H17N/c1-7(9)8-5-3-2-4-6-8/h7-8H,2-6,9H2,1H3/t7-/m0/s1

InChI key

XBWOPGDJMAJJDG-ZETCQYMHSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

125.6 °F - closed cup

Flash Point(C)

52 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service