Skip to Content
Merck
All Photos(2)

Documents

335614

Sigma-Aldrich

3-Chloropropiophenone

98%

Synonym(s):

β-Chloropropiophenone, 3-Chloro-1-phenyl-1-propanone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH2COC6H5
CAS Number:
Molecular Weight:
168.62
Beilstein:
2043580
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

113-115 °C/4 mmHg (lit.)

mp

48-50 °C (lit.)

SMILES string

ClCCC(=O)c1ccccc1

InChI

1S/C9H9ClO/c10-7-6-9(11)8-4-2-1-3-5-8/h1-5H,6-7H2

InChI key

KTJRGPZVSKWRTJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Chloropropiophenone was used in the asymmetric reduction of (S)-3-chloro-1-phenylpropanol using preheated Candida utilis cells immobilized in calcium alginate gel beads. It was also used in the synthesis of (R)-3-chloro-1-phenyl-1-propanol via asymmetric reduction using in-situ generated oxazaborolidine catalyst derived from (S)-α,α-diphenylprolinol.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric Synthesis of (R)-Fluoxetine: A Practical Approach Using Recyclable and in-situ Generated Oxazaborolidine Catalyst.
Padiya K, et al.
Chin. J. Chem., 27(6), 1137-1140 (2009)
Yang Gen-Sheng et al.
Biotechnology letters, 31(12), 1879-1883 (2009-07-28)
An efficient method for asymmetric reduction of (S)-3-chloro-1-phenylpropanol from 3-chloropropiophenone was developed using preheated Candida utilis cells immobilized in calcium alginate gel beads. Heating the immobilized cells (bead diameter 1.5 mm) at 45 degrees C for 50 min allowed the
Milada Šírová et al.
Journal of drug targeting, 25(9-10), 796-808 (2017-07-21)
Polymer carriers based on N-(2-hydroxypropyl)methacrylamide (HPMA) copolymers with incorporated organic nitrates as nitric oxide (NO) donors were designed with the aim to localise NO generation in solid tumours, thus highly increasing the enhanced permeability and retention (EPR) effect. The NO
Yu-Chang Liu et al.
Organic & biomolecular chemistry, 13(7), 2146-2152 (2014-12-23)
Styrene monooxygenase (SMO) can catalyze the kinetic resolution of secondary allylic alcohols to provide enantiopure glycidol derivatives. To overcome the low theoretical yield of kinetic resolution, we designed a one-pot two-step enzymatic cascade using prochiral α,β-unsaturated ketones as the substrates.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service