Skip to Content
Merck
All Photos(2)

Key Documents

333476

Sigma-Aldrich

Tin(IV) fluoride

Synonym(s):

Tin tetrafluoride

Sign Into View Organizational & Contract Pricing

Select a Size

2 G
$125.00

$125.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
2 G
$125.00

About This Item

Linear Formula:
SnF4
CAS Number:
Molecular Weight:
194.70
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

$125.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

Quality Level

reaction suitability

core: tin
reagent type: catalyst

SMILES string

F[Sn](F)(F)F

InChI

1S/4FH.Sn/h4*1H;/q;;;;+4/p-4

InChI key

YUOWTJMRMWQJDA-UHFFFAOYSA-J

Application

Used in toothpaste to prevent dental decay.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Shin Hum Cho et al.
The Journal of chemical physics, 152(1), 014709-014709 (2020-01-10)
A synthetic challenge in faceted metal oxide nanocrystals (NCs) is realizing tunable localized surface plasmon resonance (LSPR) near-field response in the infrared (IR). Cube-shaped nanoparticles of noble metals exhibit LSPR spectral tunability limited to visible spectral range. Here, we describe
Jian Qiu et al.
Advanced science (Weinheim, Baden-Wurttemberg, Germany), 6(1), 1800793-1800793 (2019-01-16)
Low-dimensional Ruddlesden-Popper (LDRP) lead-free perovskite has great potential due to its improved stability and oriented crystal growth, which is mainly attributed to the effective control of crystallization kinetics. However, the crystallization kinetics of LDRP lead-free perovskite films are highly limited
Edoardo Ruggeri et al.
Advanced materials (Deerfield Beach, Fla.), 31(51), e1905247-e1905247 (2019-11-12)
Halide perovskites are emerging as valid alternatives to conventional photovoltaic active materials owing to their low cost and high device performances. This material family also shows exceptional tunability of properties by varying chemical components, crystal structure, and dimensionality, providing a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service