Skip to Content
Merck
All Photos(2)

Key Documents

331384

Sigma-Aldrich

3,4,5-Trimethylphenol

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$194.00

$194.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$194.00

About This Item

Linear Formula:
(CH3)3C6H2OH
CAS Number:
Molecular Weight:
136.19
Beilstein:
1907246
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$194.00


Estimated to ship onApril 24, 2025


Request a Bulk Order

Quality Level

Assay

97%

SMILES string

Cc1cc(O)cc(C)c1C

InChI

1S/C9H12O/c1-6-4-9(10)5-7(2)8(6)3/h4-5,10H,1-3H3

InChI key

FDQQNNZKEJIHMS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Photochemical properties of 3,4,5-trimethylphenol in β-cyclodextrin complexes were studied.[1]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Photophysics and photochemistry of methylated phenols in. beta.-cyclodextrin inclusion complexes.
Monti S, et al.
The Journal of Physical Chemistry, 97(49), 13011-13016 (1993)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service