Skip to Content
Merck
All Photos(2)

Key Documents

326054

Sigma-Aldrich

Yttrium(III) carbonate hydrate

99.9% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Y2(CO3)3 · xH2O
CAS Number:
Molecular Weight:
357.84 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.9% trace metals basis
form:
powder

Assay

99.9% trace metals basis

form

powder

SMILES string

O.[Y+3].[Y+3].[O-]C([O-])=O.[O-]C([O-])=O.[O-]C([O-])=O

InChI

1S/3CH2O3.H2O.2Y/c3*2-1(3)4;;;/h3*(H2,2,3,4);1H2;;/q;;;;2*+3/p-6

InChI key

WJUZDKUDCZNBJS-UHFFFAOYSA-H

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ross K Taggart et al.
Environmental science. Processes & impacts, 20(10), 1390-1403 (2018-09-29)
Coal combustion ash is a promising alternative source of rare earth elements (REE; herein defined as the 14 stable lanthanides, yttrium, and scandium). Efforts to extract REE from coal ash will depend heavily on the location and speciation of these

Articles

Three approaches generate white light, including LED-based down-conversion for broader applications.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service