Skip to Content
Merck
All Photos(1)

Documents

325449

Sigma-Aldrich

Vinyl trifluoroacetate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3CO2CH=CH2
CAS Number:
Molecular Weight:
140.06
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

6.93 psi ( 20 °C)

Assay

98%

form

liquid

contains

10 ppm tert-butylcatechol as inhibitor

refractive index

n20/D 1.317 (lit.)

bp

40 °C/750 mmHg (lit.)

density

1.203 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

FC(F)(F)C(=O)OC=C

InChI

1S/C4H3F3O2/c1-2-9-3(8)4(5,6)7/h2H,1H2

InChI key

ZBGRMWIREQJHPK-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-25.6 °F - closed cup

Flash Point(C)

-32 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jérémie Brand et al.
Carbohydrate polymers, 169, 189-197 (2017-05-16)
Herein we propose a versatile method for the surface tailoring of cellulose nanocrystals (CNCs) based on the reactivity of vinyl ester molecules toward the accessible hydroxyl groups located at the surface of the nanoparticles. CNCs produced from wood pulp were
The ONIOM Method and Its Applications.
Lung Wa Chung et al.
Chemical reviews, 115(12), 5678-5796 (2015-04-09)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service