Skip to Content
Merck
All Photos(1)

Key Documents

322539

Sigma-Aldrich

Methyl propyl disulfide

90%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH2SSCH3
CAS Number:
Molecular Weight:
122.25
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

90%

form

liquid

impurities

~3% methyl disulfide
~6% propyl disulfide

refractive index

n20/D 1.508 (lit.)

bp

69-71 °C/43 mmHg (lit.)

density

0.98 g/mL at 25 °C (lit.)

functional group

disulfide

SMILES string

CCCSSC

InChI

1S/C4H10S2/c1-3-4-6-5-2/h3-4H2,1-2H3

InChI key

PUCHCUYBORIUSM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

109.4 °F - closed cup

Flash Point(C)

43 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Restituto Tocmo et al.
Journal of agricultural and food chemistry, 62(23), 5296-5304 (2014-05-21)
There is a growing account of the health benefits of H2S as an endogenous cell-signaling molecule. H2S from organic polysulfides, in particular, is increasingly gaining attention for their beneficial effects to cardiovascular health. Here, we studied shallot as a potential
Bofan Yu et al.
Journal of chromatography. A, 1356, 221-229 (2014-07-07)
For the expanded application area, fast trace analysis of certain high boiling point (i.e., 150-250 °C) volatile organic compounds (HVOCs) in water, a large volume-direct aqueous injection-gas chromatography (LV-DAI-GC) method was optimized for the following parameters: packed sorbent for sample

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service