Skip to Content
Merck
All Photos(1)

Key Documents

322393

Sigma-Aldrich

4-Methoxyphenyl chloroformate

98%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$116.00

$116.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$116.00

About This Item

Linear Formula:
ClCO2C6H4OCH3
CAS Number:
Molecular Weight:
186.59
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$116.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

liquid

refractive index

n20/D 1.5254 (lit.)

bp

49 °C/0.27 mmHg (lit.)

density

1.255 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

COc1ccc(OC(Cl)=O)cc1

InChI

1S/C8H7ClO3/c1-11-6-2-4-7(5-3-6)12-8(9)10/h2-5H,1H3

InChI key

CCFSGQKTSBIIHG-UHFFFAOYSA-N

Caution

Moisture sensitive material, please handle the material under inert gas (N2)/ glove bag.

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service