Skip to Content
Merck
All Photos(1)

Documents

321958

Sigma-Aldrich

Biphenylene

99%

Synonym(s):

Diphenylene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H8
CAS Number:
Molecular Weight:
152.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

mp

113-114 °C (lit.)

SMILES string

c1ccc-2c(c1)-c3ccccc-23

InChI

1S/C12H8/c1-2-6-10-9(5-1)11-7-3-4-8-12(10)11/h1-8H

InChI key

IFVTZJHWGZSXFD-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chao-Tao Tang et al.
Cellular signalling, 46, 52-63 (2018-03-03)
NADPH Oxidase 4 (NOX4), a member of the NOX family, has emerged as a significant source of reactive oxygen species, playing an important role in tumor cell proliferation, apoptosis, and other physiological processes. However, the potential function of NOX4 in
Harmanpreet Kaur et al.
Ultrasound in medicine & biology, 43(11), 2699-2712 (2017-08-16)
We evaluated the activation of mitogen-activated protein kinase (MAPK) activation through reactive oxygen species (ROS) by application of low-intensity ultrasound (LIPUS) to MC-3 T3 E1 pre-osteoblasts. The cells were subjected to one LIPUS application for either 10 or 20 min, and
Yukio Nisimoto et al.
Journal of biochemistry, 163(6), 489-501 (2018-01-25)
Nox4, a member of the NADPH- and oxygen-dependent oxidoreductases that generate reactive oxygen species (ROS), is widely expressed and constitutively active. To understand better its function and regulation, specific mutations in the Nox4 dehydrogenase (DH) domain were examined for effects
Xiaoliang Wang et al.
Journal of immunology (Baltimore, Md. : 1950), 197(10), 4090-4100 (2016-11-07)
Apoptosis is the most common form of neutrophil death under both physiological and inflammatory conditions. However, forms of nonapoptotic neutrophil death have also been observed. In the current study, we report that human neutrophils undergo necroptosis after exposure to GM-CSF
Xi-Ling Wang et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 47(2), 458-474 (2018-05-26)
Sepsis is a severe and complicated syndrome that is characterized by dysregulation of host inflammatory responses and organ failure. Cystathionine-γ-lyase (CSE)/ hydrogen sulfide (H2S) has potential anti-inflammatory activities in a variety of inflammatory diseases. NADPH oxidase 4 (Nox4), a member

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service