Skip to Content
Merck
All Photos(1)

Key Documents

318248

Sigma-Aldrich

Tetraiodoethylene

97%

Synonym(s):

1,1,2,2-Tetraiodoethene, Diiodoform, Ethylene periodide, Ethylene tetraiodide, Periodoethene, Tetraiodoethene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$425.00

$425.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$425.00

About This Item

Linear Formula:
I2C=CI2
CAS Number:
Molecular Weight:
531.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$425.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

vapor density

18.3 (vs air)

Quality Level

Assay

97%

form

solid

mp

191-193 °C (lit.)

functional group

iodo

SMILES string

I\C(I)=C(\I)I

InChI

1S/C2I4/c3-1(4)2(5)6

InChI key

ZGQURDGVBSSDNF-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Peter Stokes et al.
Rapid communications in mass spectrometry : RCM, 28(18), 2024-2030 (2014-08-19)
The atmospheric pressure solids analysis probe (ASAP) has been successfully introduced into laboratories as a routine analytical tool but the technique is labour-intensive and is not easily automated. This paper describes the modifications made to an existing atmospheric pressure gas

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service