Skip to Content
Merck
All Photos(2)

Key Documents

317551

Sigma-Aldrich

1,3-Propanediol di-p-tosylate

98%

Synonym(s):

Trimethylene glycol di-p-tosylate

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$76.70

$76.70


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$76.70

About This Item

Linear Formula:
CH3C6H4SO3(CH2)3O3SC6H4CH3
CAS Number:
Molecular Weight:
384.47
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$76.70


Estimated to ship onMay 28, 2025


Request a Bulk Order

Assay

98%

form

solid

mp

90-92 °C (lit.)

functional group

tosylate

SMILES string

Cc1ccc(cc1)S(=O)(=O)OCCCOS(=O)(=O)c2ccc(C)cc2

InChI

1S/C17H20O6S2/c1-14-4-8-16(9-5-14)24(18,19)22-12-3-13-23-25(20,21)17-10-6-15(2)7-11-17/h4-11H,3,12-13H2,1-2H3

InChI key

ODVREDGIWSDQFD-UHFFFAOYSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shivashankar Khanapur et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 58(3), 466-472 (2016-10-30)
The cerebral adenosine A
Shivashankar Khanapur et al.
Journal of medicinal chemistry, 57(15), 6765-6780 (2014-07-26)
Cerebral adenosine A2A receptors (A2ARs) are attractive therapeutic targets for the treatment of neurodegenerative and psychiatric disorders. We developed high affinity and selective compound 8 (SCH442416) analogs as in vivo probes for A2ARs using PET. We observed the A2AR-mediated accumulation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service