Skip to Content
Merck
All Photos(1)

Key Documents

316431

Sigma-Aldrich

4-Isopropyl-3-methylphenol

99%

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
$257.00

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 G
$257.00

About This Item

Linear Formula:
(CH3)2CHC6H3(CH3)OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$257.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

99%

form

crystals

mp

111-114 °C (lit.)

SMILES string

CC(C)c1ccc(O)cc1C

InChI

1S/C10H14O/c1-7(2)10-5-4-9(11)6-8(10)3/h4-7,11H,1-3H3

InChI key

IJALWSVNUBBQRA-UHFFFAOYSA-N

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nobuya Magori et al.
European journal of pharmacology, 819, 254-260 (2017-12-12)
Hinokitiol (β-thujaplicin) is a natural tropolone derivative contained in Chamaecyparis taiwanensis that has various actions including anti-inflammatory activities. Various plant-derived compounds inhibit compound action potentials (CAPs) in a manner dependent on the chemical structure of the compounds; however, the effects
Yuhe Liao et al.
Science (New York, N.Y.), 367(6484), 1385-1390 (2020-02-15)
The profitability and sustainability of future biorefineries are dependent on efficient feedstock use. Therefore, it is essential to valorize lignin when using wood. We have developed an integrated biorefinery that converts 78 weight % (wt %) of birch into xylochemicals.
Stuart J Lansdell et al.
Molecular pharmacology, 87(1), 87-95 (2014-10-24)
In common with other members of the Cys-loop family of pentameric ligand-gated ion channels, 5-hydroxytryptamine type 3 receptors (5-HT3Rs) are activated by the binding of a neurotransmitter to an extracellular orthosteric site, located at the interface of two adjacent receptor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service