Skip to Content
Merck
All Photos(3)

Key Documents

308617

Sigma-Aldrich

tert-Butyltrichlorosilane

96%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CSiCl3
CAS Number:
Molecular Weight:
191.56
Beilstein:
1736174
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

96%

form

solid

bp

132-134 °C (lit.)

mp

97-100 °C (lit.)

SMILES string

CC(C)(C)[Si](Cl)(Cl)Cl

InChI

1S/C4H9Cl3Si/c1-4(2,3)8(5,6)7/h1-3H3

InChI key

MOOUPSHQAMJMSL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

tert-Butyltrichlorosilane is a tert-alkyltrichlorosilane that can be prepared by reacting trimethyl chloride and silicon tetrachloride. It can be used in the synthesis of tert-butyl silsequioxanes by hydrolytic condensation in DMSO.[1][2]

Application

Hydrolytic condensation of tert-butyltrichlorosilane (tBuSiCl3)in DMSO and water was investigated. The process results in the formation of tBu2Si2O(OH)4, the Ti complex of the intermediate produce active heterogeneous epoxidation catalysts.[3]

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silsesquioxane-Based Homogeneous and Heterogeneous Epoxidation Catalysts Developed by Using High-Speed Experimentation
Pescarmona PP, et al.
Chemistry?A European Journal , 10(7), 1657-1665 (2004)
Silsesquioxane-Based Homogeneous and Heterogeneous Epoxidation Catalysts Developed by Using High-Speed Experimentation,
Pescarmona PP, et al.
Chemistry (Weinheim An Der Bergstrasse, Germany), 116 (38), 20433-20437 (2012)
Organomagnesium synthesis of sec-butyl-and tert-alkylchlorogermanes and their reaction with ethynylmagnesium bromide
Yarosh OG, et al.
Russ. J. Gen. Chem., 75(5), 714-718 (2005)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service