Skip to Content
Merck
All Photos(1)

Key Documents

308579

Sigma-Aldrich

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene solution

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene solution
1 of 1 reviewers received a sample product or took part in a promotion

50 wt. % in toluene

Synonym(s):

Kryptofix® 222B, Kryptand 222B

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$128.00

$128.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

Select a Size

Change View
100 MG
$128.00

About This Item

Empirical Formula (Hill Notation):
C22H36N2O6
CAS Number:
Molecular Weight:
424.53
Beilstein:
586455
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

$128.00


Estimated to ship onMay 15, 2025


Request a Bulk Order

form

liquid

Quality Level

concentration

50 wt. % in toluene

refractive index

n20/D 1.516

density

0.995 g/mL at 25 °C

functional group

ether

SMILES string

C1COCCN2CCOCCOCCN(CCO1)CCOc3ccccc3OCC2

InChI

1S/C22H36N2O6/c1-2-4-22-21(3-1)29-15-9-23-5-11-25-17-19-27-13-7-24(10-16-30-22)8-14-28-20-18-26-12-6-23/h1-4H,5-20H2

InChI key

QXLUTPDNMOEWGG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

5,6-Benzo-4,7,13,16,21,24-hexaoxa-1,10-diazabicyclo[8.8.8]hexacos-5-ene can be used as a model ligand:
  • In the metal-ligand complex formation studies to predict ligand coordination with metal ions (Eu & Gd) using magnetic resonance imaging (MRI) technique.[1]
  • In the study of coordination environment as well as redox and electronic properties of its YbII complexes.[2]

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

46.4 °F - closed cup

Flash Point(C)

8 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Screening of ligands for redox-active europium using magnetic resonance imaging
Corbin BA, et al.
Bioorganic & Medicinal Chemistry, 26(19), 5274-5279 (2018)
Spectroscopic and Electrochemical Trends in Divalent Lanthanides through Modulation of Coordination Environment
Jenks TC, et al.
Inorganic Chemistry, 59(4), 2613-2620 (2020)

Questions

Reviews

1 of 1 reviewers received a sample product or took part in a promotion

Active Filters

  1. Atlanta, GA
    • Review 1
    • Votes 3
    2 out of 5 stars.

    A bit disappointing

    I would not recommend this. I was under the impression I was receiving 100 mg of the title compound but I believe it is 100 mg of 50% w/w in toluene, so 50 mg. I was only able to recover <40 mg and some of the product in the ampoule was not toluene soluble. There is also no indication of its purity that I can find.

    Helpful?

    1. Response from MilliporeSigma:

      Thank you for taking the time to leave a review! We are sorry to hear that your experience did meet expectations. Our Technical Service team has reached out to you in effort to better understand and resolve this issue. We apologize for any inconvenience this may have caused.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service