Skip to Content
Merck
All Photos(1)

Documents

308439

Sigma-Aldrich

Selenic acid

94%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2SeO4
CAS Number:
Molecular Weight:
144.97
EC Number:
MDL number:
UNSPSC Code:
12352300

Assay

94%

refractive index

n20/D 1.5174 (lit.)

density

2.511 g/mL at 25 °C (lit.)

SMILES string

O[Se](O)(=O)=O

Looking for similar products? Visit Product Comparison Guide

Packaging

Packaged in glass bottles

replaced by


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service